Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Collect, transport and process capillary blood. Greiner BioOne™ MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
Brand:  Greiner Bio-One 450477
Product Code. 12669092
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)