missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MiniCollect™ Capillary Blood Collection System Accessories

Product Code. 13058602
Change view
Click to view available options
Product Type:
EDTA Tube
Tube Size:
80 μL
Color:
Lavender
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Tube Size Color
13058602 EDTA Tube 80 μL Lavender
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13058602 Supplier Greiner Bio-One Supplier No. 450434
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with MiniCollect Capillary Blood Collection System

  • Capillary or funnel can be inserted into MiniCollect tubes without removing the cap
  • 13 × 75mm carrier tubes facilitate bar code labeling, centrifugation, and storage
  • Use amber carrier tube for sheilding light-sensitive analytes

TRUSTED_SUSTAINABILITY

Specifications

Color Lavender
Product Type EDTA Tube
Capacity (Metric) 80 μL
For Use With (Equipment) Greiner Vacuette MiniCollect Capillary Blood Collection System
Tube Size 80 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.