missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Luer Adapter

Product Code. 15432510
Change view
Click to view available options
Product Type:
Luer Adapter
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15432510 Luer Adapter Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15432510 Supplier Greiner Bio-One Supplier No. 450070
  • ONLINE EXCLUSIVE PRICE
    £1500.00
    £1374.04 / Pack of 100
    Save £125.96
    8% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Safety/Blood Collection sets or Luer needles in routine blood collection

  • Sterile, single-use device
  • Manufactured from stainless steel and polypropylene
  • Fitted with a safety valve at one end
  • Perforated label simplifies identification and serves as a seal of integrity

TRUSTED_SUSTAINABILITY

Specifications

Product Type Luer Adapter
Material Stainless Steel, Polypropylene
Sterility Sterile
Certifications/Compliance CE, OSHA
Disposable Single-use
For Use With (Equipment) Safety/Blood collection sets or 20G luer needles in routine Blood collection
Includes Safety Valve at one end
Storage Requirements 4°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.