missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Green'R Floor Cleaner

Product Code. 17830255
Change view
Click to view available options
Format:
Bottle
Can
Quantity:
2 x 5 L
250 x 20 mL
Unit Size:
10 L
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Quantity unitSize
17830255 Bottle 250 x 20 mL Pack of 250
17810275 Can 2 x 5 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17830255 Supplier Christeyns™ Supplier No. 547509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Green'R Floor is a very efficient, non-foaming multipurpose cleaner that shines without leaving streaks.

  • Diluted in manual use or in a scrubber
  • Eco-labelled neutral PH detergent
  • Non-foaming and approved for food contact
  • Ensures quick and effective cleaning of all floors and all washable surfaces
  • Suitable for regular maintenance
  • Floral scent

Specifications

Product Type Neutral Detergent
Color-coded No
Form Concentrated Liquid
pH Neutral
Product Line Cleanware
Scent Floral
Sterility Non-sterile
Format Bottle
Quantity 250 x 20 mL
compliance-icons
Product Identifier
  • GreenR Floors
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Phosphonobutane-1,2,4-tricarboxylic acid, Isotridecanol, ethoxylated, sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.