missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Green'R Floor Cleaner
Description
- Diluted in manual use or in a scrubber
- Eco-labelled neutral PH detergent
- Non-foaming and approved for food contact
- Ensures quick and effective cleaning of all floors and all washable surfaces
- Suitable for regular maintenance
- Floral scent
Specifications
Specifications
| Product Type | Neutral Detergent |
| Color-coded | No |
| Form | Concentrated Liquid |
| pH | Neutral |
| Product Line | Cleanware |
| Scent | Floral |
| Sterility | Non-sterile |
| Format | Can |
| Quantity | 2 x 5 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?