Learn More
Tenak™ Green Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand:  Tenak™ TE22003
Additional Details : Weight : 0.08000kg
 View more versions of this product
Product Code. 11577294
  • £80.15 / Pack of 36
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Specifications
Green
Square
133 x 133 x 32 mm
Cardboard
Cryo Box