missing translation for 'onlineSavingsMsg'
Learn More

GRB7 Antibody (OTI6H2) [PE], Novus Biologicals™

Product Code. 30687795 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687795 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687795 Supplier Novus Biologicals Supplier No. NBP344728PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GRB7 Monoclonal antibody specifically detects GRB7 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB7
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI6H2
Conjugate PE
Formulation PBS
Gene Alias B47, Epidermal growth factor receptor GRB-7, GRB7 adapter protein, growth factor receptor-bound protein 7
Host Species Mouse
Immunogen Full length human recombinant protein of human GRB7 (NP_005301) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 2886
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.