missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRB2 Recombinant Rabbit Monoclonal Antibody (9K6H0)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535238
This item is not returnable.
View return policy
Description
Immunogen sequence: YFLWVVKFNS LNELVDYHRS TSVSRNQQIF LRDIEQVPQQ PTYVQALFDF DPQEDGELGF RRGDFIHVMD NSDPNWWKGA CHGQTGMFPR NYVTPVNRNV.
GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2-Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.
Specifications
| GRB2 | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62993, P62994, Q60631 | |
| GRB2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 9K6H0 | |
| Unconjugated | |
| GRB2 | |
| AA408164; abundant SRC homology; adapter protein GRB2; Adapter protein GRB2-A; ASH; ASH protein; EGFRBP-GRB2; epidermal growth factor receptor-binding protein GRB2; grb2; grb2.L; grb2-a; grb2-b; Grb3-3; growth factor receptor bound protein 2; growth factor receptor bound protein 2 L homeolog; growth factor receptor-bound protein 2; Growth factor receptor-bound protein 2-A; growth factor receptor-bound protein 3; HT027; hypothetical protein LOC535298; MST084; MSTP084; NCKAP2; OTTHUMP00000166097; OTTHUMP00000166098; protein Ash; SH2/SH3 adapter GRB2; SH2/SH3 adapter GRB2-A; SH2/SH3 adaptor Grb2; XELAEV_18044055mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14784, 2885, 81504 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction