missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRB14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528914
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (88%), Rat (86%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. This protein likely has an inhibitory effect on receptor tyrosine kinase signaling and, in particular, on insulin receptor signaling. This gene may play a role in signaling pathways that regulate growth and metabolism. Transcript variants have been reported for this gene, but their full-length natures have not been determined to date.
Specifications
| GRB14 | |
| Polyclonal | |
| Unconjugated | |
| GRB14 | |
| AI505286; GRB14; GRB14 adapter protein; growth factor receptor bound protein 14; growth factor receptor-bound protein 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2888, 50915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.43 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300 | |
| Q14449, Q9JLM9 | |
| GRB14 | |
| Recombinant protein encompassing a sequence within the center region of human GRB14. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction