missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23591-1-AP-150UL
This item is not returnable.
View return policy
Description
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.Specifications
| GRB10 | |
| Polyclonal | |
| Unconjugated | |
| GRB10 | |
| Grb 10, GRB IR, GRB10, GRB10 adapter protein, GRBIR, IRBP, KIAA0207, MEG1, RSS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14783, 2887, 498416 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0CE43, Q13322, Q60760 | |
| GRB10 | |
| GRB10 Fusion Protein Ag20355 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction