missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB10 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41159F
This item is not returnable.
View return policy
Description
GRB10 Polyclonal antibody specifically detects GRB10 in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence
Specifications
| GRB10 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunofluorescence | |
| FITC | |
| Grb-10, GRB10 adapter protein, GRB10 adaptor protein, GRBIR, GRB-IR, growth factor receptor-bound protein 10, Insulin receptor-binding protein Grb-IR, IRBP, KIAA0207, maternally expressed gene 1, MEG1, RSS | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human GRB10. The immunogen is located within amino acids 440 - 490 of GRB10. Amino Acid Squence: EEGHAWRKRSTRMNILG | |
| 0.1 mL | |
| Cancer | |
| 2887 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction