missing translation for 'onlineSavingsMsg'
Learn More

GRB10 Antibody (1A7), Novus Biologicals™

Product Code. 18396388 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396388 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396388 Supplier Novus Biologicals Supplier No. H00002887M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GRB10 Monoclonal antibody specifically detects GRB10 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB10
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH24285
Gene Alias Grb-10, GRB10 adapter protein, GRB10 adaptor protein, GRBIR, GRB-IR, growth factor receptor-bound protein 10, Insulin receptor-binding protein Grb-IR, IRBP, KIAA0207, maternally expressed gene 1, MEG1, RSS
Host Species Mouse
Immunogen GRB10 (AAH24285, 61 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AVRRLQEEDQQFRTSSLPAIPNPFPELCGPGSPPVLTPGSLPPSQAAAKQDVKVFSEDGTSKVVEILADMTARDLCQLLVYKSHCVDDNS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2887
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.