Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
Brand:  Deutsch Neumann™ 1010121
Additional Details : Weight : 0.01530kg
 View more versions of this product
Product Code. 10169890
  • £1.15 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
Gray
30mm
27 mm
Natural Rubber
Tapered Stopper
21 mm
1/Ea.