Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
Brand: Deutsch Neumann™ 1010164
Additional Details : Weight : 0.23400kg
Description
- Acc. to DIN 12 871
- Durometer 40 Shore A
- Available in a wide range of sizes
Specifications
Gray | |
55mm | |
75.5 mm | |
Natural Rubber |
Tapered Stopper | |
64.5 mm | |
1/Ea. |