Learn More
Manutan Gravity roller bearing conveyor, 70kg steel roller bearing capacity, width 600mm, pitch 150mm
Product Code. 12841479
Encompass_Preferred
Product Code. 12841479 Supplier Manutan Supplier No. A341747
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Conveyor tracks manufactured from high quality 2mm thick steel and supplied complete with zinc plated steel or high impact PVC roller bearings. Height adjustable supports allow track to decline for smooth movement. Low maintenance for trouble free service. Track sides are pre-punched every 150mm to accept support stands and guard rails.
  • Roller bearing pitch: 150mm
  • Track width: 600mm