Learn More
Manutan Gravity roller bearing conveyor, 20kg PVC roller bearing capacity, width 450mm, roller bearing pitch 150mm
Product Code. 12824899
Encompass_Preferred
Product Code. 12824899
Brand: Manutan A341392
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Conveyor tracks manufactured from high quality 2mm thick steel and supplied complete with zinc plated steel or high impact PVC roller bearings. Height adjustable supports allow track to decline for smooth movement. Low maintenance for trouble free service. Track sides are pre-punched every 150mm to accept support stands and guard rails.
  • Per metre
  • Roller bearing pitch: 150mm