missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP65 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36527DL594
This item is not returnable.
View return policy
Description
GRASP65 Polyclonal antibody specifically detects GRASP65 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GRASP65 | |
| Polyclonal | |
| Western Blot | |
| Golgi peripheral membrane protein p65, Golgi phosphoprotein 5MGC118897, golgi reassembly stacking protein 1, 65kDa, Golgi reassembly-stacking protein 1, Golgi reassembly-stacking protein of 65 kDa, GRASP65MGC118894, P6565 kDa | |
| The immunogen recognized by this antibody maps to a region between residue 390 to 440 of human Golgi Reassembly Stacking Protein 1, 65kDa using the numbering given in entry NP_114105.1. | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 64689 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction