missing translation for 'onlineSavingsMsg'
Learn More

GRASP55 Antibody, Novus Biologicals™

Product Code. 18369339 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369339 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369339 Supplier Novus Biologicals Supplier No. H00026003B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GRASP55 Polyclonal antibody specifically detects GRASP55 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP55
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_056345.3
Gene Alias Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59
Host Species Mouse
Immunogen GORASP2 (NP_056345.3, 1 a.a. - 452 a.a.) full-length human protein. MGSSQSVEIPGGGTEGYHVLRVQENSPGHRAGLEPFFDFIVSINGSRLNKDNDTLKDLLKANVEKPVKMLIYSSKTLELRETSVTPSNLWGGQGLLGVSIRFCSFDGANENVWHVLEVESNSPAALAGLRPHSDYIIGADTVMNESEDLFSLIETHEAKPLKLYVYNTDTDNCREVIITPNSAWGGEGSLGCGIGYGYLHRIPTRPFEEGKKISLPGQMAGTPITPLKDGFTEVQLSSVNPPSLSPPGTTGIEQSLTGLSISSTPPAVSSVLSTGVPTVPLLPPQVNQSLTSVPPMNPATTLPGLMPLPAGLPNLPNLNLNLPAPHIMPGVGLPELVNPGLPPLPSMPPRNLPGIAPLPLPSEFLPSFPLVPESSSAASSGELLSSLPPTSNAPSDPATTTAKADAASSLTVDVTPPTAKAPTTVEDRVGDSTPVSEKPVSAAVDANASESP
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 26003
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.