missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP55 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£280.00 - £452.00
Specifications
| Antigen | GRASP55 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18431131
|
Novus Biologicals
NBP1-89747-25ul |
25 μL |
£280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761674
|
Novus Biologicals
NBP1-89747 |
£452.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
GRASP55 Polyclonal specifically detects GRASP55 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GRASP55 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9H8Y8 | |
| 26003 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PTRPFEEGKKISLPGQMAGTPITPLKDGFTEVQLSSVNPPSLSPPGTTGIEQSLTGLSISSTPPAVSSVLSTGV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Golgi Apparatus Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59 | |
| GORASP2 | |
| IgG | |
| Affinity Purified | |
| Specificity of human GRASP55 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title