missing translation for 'onlineSavingsMsg'
Learn More

GRASP55 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18379252 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379252 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379252 Supplier Novus Biologicals Supplier No. NBP299119G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GRASP55 Polyclonal specifically detects GRASP55 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP55
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human GRASP55.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 26003
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.