missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP55 Antibody (CL2610) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36779R
This item is not returnable.
View return policy
Description
GRASP55 Monoclonal antibody specifically detects GRASP55 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| GRASP55 | |
| Monoclonal | |
| DyLight 550 | |
| Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59 | |
| This antibody was developed using a recombinant protein derived from Q9H8Y8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 26003 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL2610 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction