missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRASP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559242
This item is not returnable.
View return policy
Description
Immunogen sequence: VEMVTFVCRV HESSPAQLAG LTPGDTIASV NGLNVEGIRH REIVDIIKAS GNVLRLETLY GTSIRKAELE ARLQYLKQTL YEKWG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GRASP (General receptor for phosphoinositides 1-associated scaffold protein) plays a role in intracellular trafficking and contributes to the macromolecular organization of group 1 metabotropic glutamate receptors (mGluRs) at synapses, linking receptors to neuronal proteins.This protein contains conserved domains, including a leucine zipper sequence, PDZ domain and a C-terminal PDZ-binding motif.
Specifications
| GRASP | |
| Polyclonal | |
| Unconjugated | |
| GRASP | |
| 95 kDa postsynaptic density protein discs-large ZO-1 domain-containing protein; general receptor for phosphoinositides 1 associated scaffold protein; general receptor for phosphoinositides 1-associated scaffold protein; grasp; GRASP-1; graspl; GRIP1; GRIP-1; GRIPAP1; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein like; Grp1 (general receptor for phosphoinositides)-associated scaffold protein; GRP1-associated scaffold protein; hypothetical protein LOC558950; KIAA1167; LOW QUALITY PROTEIN: general receptor for phosphoinositides 1-associated scaffold protein; MNCb-4428; protein TAMALIN; PSD-95 PDZ domain-containing protein; Tamalin; trafficking regulator and scaffold protein tamalin; zgc:136876 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 160622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z6J2 | |
| GRASP | |
| Recombinant protein corresponding to Human GRASP1. Recombinant protein control fragment (Product #RP-97839). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction