missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRASP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566448

Product Code. 15999144

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PADELYAALED YHPAELYRAL AVSGGTLPRR KGSGFRWKNL SQSPEQQRKV LTLEKED Highest antigen sequence identity to the following orthologs - mouse 91%, rat 93%.

GRASP (General receptor for phosphoinositides 1-associated scaffold protein) plays a role in intracellular trafficking and contributes to the macromolecular organization of group 1 metabotropic glutamate receptors (mGluRs) at synapses, linking receptors to neuronal proteins.This protein contains conserved domains, including a leucine zipper sequence, PDZ domain and a C-terminal PDZ-binding motif.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRASP
Polyclonal
Unconjugated
GRASP
95 kDa postsynaptic density protein discs-large ZO-1 domain-containing protein; general receptor for phosphoinositides 1 associated scaffold protein; general receptor for phosphoinositides 1-associated scaffold protein; grasp; GRASP-1; graspl; GRIP1; GRIP-1; GRIPAP1; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein like; Grp1 (general receptor for phosphoinositides)-associated scaffold protein; GRP1-associated scaffold protein; hypothetical protein LOC558950; KIAA1167; LOW QUALITY PROTEIN: general receptor for phosphoinositides 1-associated scaffold protein; MNCb-4428; protein TAMALIN; PSD-95 PDZ domain-containing protein; Tamalin; trafficking regulator and scaffold protein tamalin; zgc:136876
Rabbit
Antigen Affinity Chromatography
RUO
160622
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7Z6J2
GRASP
Recombinant Human GRASP. Recombinant protein control fragment (Product #RP-107123).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.