missing translation for 'onlineSavingsMsg'
Learn More

Grapiprant, MedChemExpress

Product Code. 16499717
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499717

Brand: Medchem Express HY16781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Grapiprant (CJ-023423) is a selective EP4 receptor antagonist whose physiological ligand is prostaglandin E2 (PGE2). Grapiprant displaces [3H]-PGE2 (1 nM) binding to dog recombinant EP4 receptor with IC50 value of 35 nM and Ki value of 24 nM. Grapiprant has the potential for osteoarthritic pain and inflammation treatment[3] .

Grapiprant (CJ-023423) is a selective EP4 receptor antagonist whose physiological ligand is prostaglandin E2 (PGE2). Grapiprant displaces [3H]-PGE2 (1 nM) binding to dog recombinant EP4 receptor with IC50 value of 35 nM and Ki value of 24 nM. Grapiprant has the potential for osteoarthritic pain and inflammation treatment[3] .

Specifications

Chemical Name or Material Grapiprant
CAS 415903-37-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H29N5O3S
Quantity 5 mg
Synonym CJ-023423 RQ-00000007 AAT-007
Solubility Information DMSO : ≥ 50 mg/mL (101.71 mM)
SMILES O=S(C1=CC=C(C)C=C1)(NC(NCCC2=CC=C(N3C(CC)=NC4=C3C=C(C)N=C4C)C=C2)=O)=O
Molecular Weight (g/mol) 491.61
Formula Weight 491.61
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.