Learn More
Trajan Scientific and Medical™ Graphite Vespel™ Ferrules for Shimadzu
Product Code. 10454302
Change view
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
0.32 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside unitSize
10454302 0.10 to 0.25 mm Pack of 10
10592203 0.32 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10454302 Supplier Trajan Scientific and Medical™ Supplier No. 0726561
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Provides leak-free system connections. Trajan Scientific and Medical Graphite Vespel™ Ferrules for Shimadzu leads to improved chromatography for GC systems.
  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column
Specifications
Type Graphite Vespel Ferrule for 0.1-0.25mm ID Columns for Shimadzu MS
For Use With (Equipment) QP5000-II and QP5050
Material 15% Graphite, 85% Vespel
Diameter (Metric) Inside 0.10 to 0.25 mm