missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Graphite GC Liner O-Rings

Graphite liner O-rings for use with GC liners.

Brand:  Trajan™ 0726006

Code : 13

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10370841

  • £40.90 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

  • Graphite Sealing Rings
  • Temperature stable up to 450°C
  • Supplied in packs of 10
Specifications

Specifications

Agilent Technologies 2.0 mm ID Splitless (Quartz), Straight-Through Liner
Graphite
Sealing Ring
For Use with Liners
450°C
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers