missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRAP2 Monoclonal Antibody (UW40)

Product Code. 12372353
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12372353 100 μg Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12372353 Supplier Invitrogen™ Supplier No. MA110341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes GRAP2/GADS, a 41 kDa cytoplasmic adaptor protein that plays a pivotal role during the early events of signal transduction in T cells.

This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRAP2
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone UW40
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 15 mM sodium azide; pH 7.4
Gene Grap2
Gene Accession No. O75791
Gene Alias adapter protein GRID; Gads; GADS protein; Grap2; GRAP-2; GRB2L; GRB-2-like protein; GRB2-related adapter protein 2; Grb2-related adaptor downstream of Sch; GRB2-related adaptor protein 2; GRB-2-related monocytic adapter protein; GRB2-related protein with insert domain; GRBLG; GRBX; Grf40; grf-40; Grf40 adapter protein; GRID; Growth factor receptor-binding protein; growth factor receptor-bound protein 2-related adaptor protein 2; GrpL; Hematopoietic cell-associated adapter protein GrpL; hematopoietic cell-associated adaptor protein GRPL; MONA; monocytic adapter; monocytic adaptor; P38; Protein GADS; RP3-370M22.1; SH3-SH2-SH3 adapter Mona; SH3-SH2-SH3 adaptor molecule
Gene Symbols Grap2
Host Species Mouse
Immunogen GST-fusion human GRAP2/GADS protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9402
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.