missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599632
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Grap.
This gene encodes a member of the GRB2/Sem5/Drk family. This member functions as a cytoplasmic signaling protein which contains an SH2 domain flanked by two SH3 domains. The SH2 domain interacts with ligand-activated receptors for stem cell factor and erythropoietin, and facilitates the formation of a stable complex with the BCR-ABL oncoprotein. This protein also associates with the Ras guanine nucleotide exchange factor SOS1 (son of sevenless homolog 1) through its N-terminal SH3 domain.
Specifications
| GRAP | |
| Polyclonal | |
| Unconjugated | |
| GRAP | |
| 8430435N19Rik; BB233594; GRAP; GRB2 related adaptor protein; GRB2-related adapter protein; GRB2-related adaptor protein; growth factor receptor-bound protein 2-related adaptor protein; MGC64880 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10750, 71520 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13588, Q9CX99 | |
| GRAP | |
| A synthesized peptide derived from human GRAP(Accession Q13588), corresponding to amino acid residues I185-L217. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction