missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111238
This item is not returnable.
View return policy
Description
Immunogen sequence: ELVDFYRTTT IAKKRQIFLR DEEPLLKSPG ACFAQAQFDF SAQDPSQLSF RRGDIIEVLE RPDPHWWRGR SCGRVGFFPR SYVQPVHL Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 84%.
This gene encodes a member of the GRB2/Sem5/Drk family. This member functions as a cytoplasmic signaling protein which contains an SH2 domain flanked by two SH3 domains. The SH2 domain interacts with ligand-activated receptors for stem cell factor and erythropoietin, and facilitates the formation of a stable complex with the BCR-ABL oncoprotein. This protein also associates with the Ras guanine nucleotide exchange factor SOS1 (son of sevenless homolog 1) through its N-terminal SH3 domain.
Specifications
| GRAP | |
| Polyclonal | |
| Unconjugated | |
| GRAP | |
| 8430435N19Rik; BB233594; GRAP; GRB2 related adaptor protein; GRB2-related adapter protein; GRB2-related adaptor protein; growth factor receptor-bound protein 2-related adaptor protein; MGC64880 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13588 | |
| GRAP | |
| Recombinant Protein corresonding to Human GRAP. Recombinant protein control fragment (Product #RP-96521). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction