missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35542-20ul
This item is not returnable.
View return policy
Description
GRAP Polyclonal antibody specifically detects GRAP in Human samples. It is validated for ELISA,Western Blot
Specifications
| GRAP | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| GRB2-related adapter protein, GRB2-related adaptor protein, growth factor receptor-bound protein 2-related adaptor protein, MGC64880 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 128-217 of human GRAP (NP_006604.1).,, Sequence:, LNELVDFYRTTTIAKKRQIFLRDEEPLLKSPGACFAQAQFDFSAQDPSQLSFRRGDIIEVLERPDPHWWRGRSCGRVGFFPRSYVQPVHL | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10750 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction