missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme K Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£236.00 - £553.00
Specifications
| Antigen | Granzyme K |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18324664
|
Novus Biologicals
NBP3-17052-25UL |
25 μg |
£236.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375016
|
Novus Biologicals
NBP3-17052-100UL |
100 μg |
£553.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granzyme K Polyclonal antibody specifically detects Granzyme K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Granzyme K | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Immunology | |
| PBS, pH 7.2, 40% glycerol | |
| 3003 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, granzyme 3, granzyme K, granzyme K (granzyme 3; tryptase II), granzyme K (serine protease, granzyme 3; tryptase II), Granzyme-3, NK-Tryp-2, NK-tryptase-2, PRSS, TRYP2granzyme-3, tryptase II | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LSKNEASKQTLEIKKFIPFSRVTSDPQSNDIMLVKLQTAAKLNKHVKMLHIRSKTSLRSGTK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title