missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granzyme K Monoclonal Antibody (C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551116
This item is not returnable.
View return policy
Description
Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.
Specifications
| Granzyme K | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| O35205, P49864 | |
| GZMK | |
| Recombinant Mouse GZMK protein, Gln44-Val227 (Accession #O35205). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C8 | |
| Unconjugated | |
| GZMK | |
| Fragmentin-3; granzyme 3; granzyme K; granzyme K (granzyme 3; tryptase II); granzyme K (serine protease, granzyme 3; tryptase II); granzyme-3; GrK; GZMK; NK-Tryp-2; NK-tryptase-2; RNK-Tryp-2; TRYP2; tryptase II | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 14945, 29165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction