missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567564
Additional Details : Weight : 0.01000kg
Description
Granzyme D is thought to be necessary for target cell lysis in cell-mediated immune responses.Specifications
Granzyme D | |
Polyclonal | |
Unconjugated | |
Gzmd | |
CCP5; Ctla5; Ctla-5; Granzyme D; Gzmd | |
Rabbit | |
affinity chromatography | |
RUO | |
14941 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P11033 | |
Gzmd | |
Synthetic peptide derived from mouse Granzyme B. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |