missing translation for 'onlineSavingsMsg'
Learn More

Granzyme D Antibody (RM0246-8G14), PerCP, Novus Biologicals™

Product Code. 18408671 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408671 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408671 Supplier Novus Biologicals Supplier No. NBP212409PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Granzyme D Monoclonal antibody specifically detects Granzyme D in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme D
Applications Western Blot
Classification Monoclonal
Clone RM0246-8G14
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CCP5, Ctla5, Ctla-5, granzyme D
Host Species Rat
Immunogen Mouse recombinant Granzyme D
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14941
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.