Learn More
Granzyme B Mouse anti-Human, Alexa Fluor 700, Clone: GB11, BD
Mouse Monoclonal Antibody
£271.00 - £382.00
Specifications
| Antigen | Granzyme B |
|---|---|
| Clone | GB11 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
The GB11 antibody specifically reacts with human granzyme B, a serine protease of approximately 32 kDa. Granzyme B is stored in the granules of cytotoxic T lymphocytes and NK cells along with the pore-forming protein perforin. In the classic model of target cell lysis, perforins create holes in the target cell membrane allowing entrance of granzymes. Granzyme B has been shown to act on specific substrates including caspase-3, -7, -9, and -10 which in turn give rise to enzymes that mediate apoptosis. Granzyme B may also be involved in the hydrolysis of extracellular matrix components. Detectable levels of granzyme B have been detected in sera from healthy volunteers. The immunogen used to generate the GB11 hybridoma was human granzyme B isolated from an NK cell line.
Specifications
| Granzyme B | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Granzyme B | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| GB11 | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| RUO | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| GZMB; Granzyme-2; CCPI; CGL1; CSPB; CTLA1; CTSGL1; GRB; HLP; SECT |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.