missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody (CL10662) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07984AF647
This item is not returnable.
View return policy
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Granzyme B | |
| Monoclonal | |
| Alexa Fluor 647 | |
| C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
| This antibody was developed using a recombinant protein derived from P10144, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis | |
| 3002 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10662 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction