Learn More
Epredia™ Lab Vision™ Granzyme B Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Granzyme B Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1157-RQ
Description
Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified. One of these, human granzyme B, is involved in target cell apoptosis during lymphocyte-mediated cytotoxicity. Exocytosis of granzyme-containing granules in the cytoplasm of the target cell leads to the induction of DNA fragmentation and apoptosis.Host Species: Mouse
Clone: GZB01
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein encoding the N-terminus of the mature granzyme B
Molecular Weight: 32 kDa
Positive Control: Hodgkin's lymphoma, Tonsil
Cellular Localization: Granular cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Specifications
| Granzyme B Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein encoding the N-terminus of the mature granzyme B | |
| IVD | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| GZB01 | |
| Mouse | |
| 12 mL | |
| Apoptosis | |
| Ready-to-Use |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.