missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11288-1-AP-150UL
This item is not returnable.
View return policy
Description
Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface ′nonself′ antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells.Specifications
| Granzyme A | |
| Polyclonal | |
| Unconjugated | |
| GZMA | |
| CTL tryptase, CTLA3, Fragmentin 1, Granzyme 1, Granzyme A, GZMA, H factor, Hanukkah factor, HF, HFSP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14938, 3001 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11032, P12544 | |
| Gzma | |
| Granzyme A Fusion Protein Ag1818 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction