missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Granzyme A Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £400.00
Specifications
| Antigen | Granzyme A |
|---|---|
| Clone | 356422 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
18717719
![]() |
R&D Systems
MAB2905-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18170421
|
R&D Systems
MAB2905 |
100 μg |
£400.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Granzyme A Monoclonal specifically detects Granzyme A in Human samples. It is validated for Western Blot.Specifica
| Granzyme A | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P12544 | |
| 3001 | |
| Mouse myeloma cell line NS0-derived recombinant human Granzyme A Cys26-Val262 Accession # P12544 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Granzyme A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Granzyme B, rhGranzyme H, recombinant mouse (rm) Granzyme D, or rmGranzyme G is observed. |
| 356422 | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CTL tryptase, CTLA3, CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factorserine protease), Cytotoxic T-lymphocyte proteinase 1, EC 3.4.21, fragmentin-1, granzyme A, granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3), Granzyme-1, GZMA, H factor, Hanukkah factor, HF, HFSP, HFSPEC 3.4.21.78 | |
| GZMA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto