Learn More
Granzyme A Monoclonal Antibody (GzA-3G8.5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-5831-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This Gza-3G8.5 monoclonal antibody reacts with mouse Granzyme A. Granzyme A is the most abundantly expressed of the ten granzyme serine proteases that have been identified in mice. Granzymes are proteins released from the granules of NK cells and cytotoxic T lymphocytes that induce death in target cells by cleavage of intracellular substrates and play a critical role in immune defense against viruses, tumors, and intracellular bacteria. Granzyme A activates caspase-independent cell death pathways morphologically similar to apoptosis and characterized by mitochondrial and DNA damage. It may also play a role in inflammation, as the precursor form of IL-1 beta (pro-IL-1 beta) is among its target substrates. Granzyme A shares overlapping substrate specificity with the closely-related Granzyme K, which is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTLs. Applications Reported: This GzA-3G8.5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This GzA-3G8.5 antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface ′nonself′ antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells.Specifications
Granzyme A | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P11032 | |
Gzma | |
Affinity chromatography | |
RUO | |
14938 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
GzA-3G8.5 | |
PerCP-eFluor 710 | |
Gzma | |
CTLA3, HFSP | |
Mouse | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
For Research Use Only.