missing translation for 'onlineSavingsMsg'
Learn More

Granulysin Mouse anti-Human, Clone: B-R32, Novus Biologicals™

Product Code. 18330045 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330045 200 μg 200µL
18358931 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18330045 Supplier Novus Biologicals Supplier No. NBP318105200UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulysin
Applications ELISA
Classification Monoclonal
Clone B-R32
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias D2S69Elymphocyte-activation gene 2, granulysin, LAG2, LAG-2, Lymphokine LAG-2, NKG5519, Protein NKG5, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519
Host Species Mouse
Immunogen Recombinant human Granulysin (NKG5 and LAG-2). Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10578
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.