missing translation for 'onlineSavingsMsg'
Learn More

Granulysin Antibody (786314) [DyLight 550], Novus Biologicals™

Product Code. 30733676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733676 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733676 Supplier Novus Biologicals Supplier No. FAB31381L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulysin
Applications ELISA
Classification Monoclonal
Clone 786314
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias D2S69Elymphocyte-activation gene 2, granulysin, LAG2, LAG-2, Lymphokine LAG-2, NKG5519, Protein NKG5, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Granulysin, Arg23-Leu145, Accession # P22749
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10578
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.