Learn More
Invitrogen™ Granulins Recombinant Superclonal™ Antibody (2HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710191
Description
This antibody is predicted to react with equine, mouse, rabbit and rat based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
PCDGF is a glycoprotein and growth factor involved in the regulation of mitosis, early embryonic development, cell division, cell migration, cell survival, and hypothalamic differentiation. PCDGF is also known as progranulin because it is the precursor for smaller proteins known as epithelins or granulins.
Specifications
| Granulins | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P28799 | |
| GRN | |
| Peptide corresponding to amino acids 573-588 of human PC cell-derived growth factor. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 2HCLC | |
| Unconjugated | |
| GRN | |
| acrogranin; acrogranulin; CLN11; epithelin; Epithelin precursor; Epithelin/granulin precursor; Epithelin-1; Epithelin-2; GEP; Glycoprotein of 88 Kda; GP88; granulin; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; granulins; Grn; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; progranulin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2896 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.