Learn More
Granulin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18410-1-AP-20UL
Description
The Granulin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Granulin. This antibody recognizes human, mouse, rat antigen. The Granulin antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| Granulin | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2896 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |