missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulin Rabbit anti-Human, Clone: 4X3I7, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Granulin |
|---|---|
| Clone | 4X3I7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18324935
|
Novus Biologicals
NBP3-16566-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394536
|
Novus Biologicals
NBP3-16566-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Western BlotSpecifications
| Granulin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2896 | |
| IgG | |
| Affinity purified |
| 4X3I7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human Granulin (P28799). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title