missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulin Antibody (OTI3H6), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70847C
This item is not returnable.
View return policy
Description
Granulin Monoclonal specifically detects Granulin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Granulin | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GRN | |
| Recombinant protein fragment corresponding to amino acids 327-593 of human Granulin (NP_002078) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3H6 | |
| Western Blot, Immunohistochemistry | |
| acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 2896.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction