missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulin Antibody (OTI3H6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70847CL1
This item is not returnable.
View return policy
Description
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Granulin | |
| Monoclonal | |
| CoraFluor 1 | |
| acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
| Human recombinant protein fragment corresponding to amino acids 327-593 of human GRN(NP_002078) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Lipid and Metabolism | |
| 2896 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI3H6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction