missing translation for 'onlineSavingsMsg'
Learn More

Granisetron Hydrochloride, MedChemExpress

Product Code. p-200096405
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16494217

Brand: Medchem Express HYB0071A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy.

Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy.

Specifications

Chemical Name or Material Granisetron Hydrochloride
CAS 107007-99-8
Color White
Purity Grade Notes Research
Molecular Formula C18H25ClN4O
Quantity 50 mg
Synonym BRL 43694A
Solubility Information H2O : 100 mg/mL (286.64 mM; Need ultrasonic) ∣DMSO : 7.69 mg/mL (22.04 mM; Need ultrasonic)
SMILES CN1N=C(C(N[C@H]2C[C@H](N3C)CCC[C@H]3C2)=O)C4=CC=CC=C41.[H]Cl
Molecular Weight (g/mol) 348.87
Formula Weight 348.87
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.