missing translation for 'onlineSavingsMsg'
Learn More

Granisetron Hydrochloride, MedChemExpress

Product Code. 16494217
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494217 50 mg 50mg
16448076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16494217 Supplier Medchem Express Supplier No. HYB0071A
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy.

Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy.

Specifications

Chemical Name or Material Granisetron Hydrochloride
CAS 107007-99-8
Color White
Purity Grade Notes Research
Molecular Formula C18H25ClN4O
Quantity 50 mg
Synonym BRL 43694A
Solubility Information H2O : 100 mg/mL (286.64 mM; Need ultrasonic) ∣DMSO : 7.69 mg/mL (22.04 mM; Need ultrasonic)
SMILES CN1N=C(C(N[C@H]2C[C@H](N3C)CCC[C@H]3C2)=O)C4=CC=CC=C41.[H]Cl
Molecular Weight (g/mol) 348.87
Formula Weight 348.87
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.