missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Granisetron hydrochloride

Product Code. 18752794
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752794 50 mg 50mg
18702514 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752794 Supplier R&D Systems™ Supplier No. 2903/50
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT3 antagonist

5-HT3 receptor antagonist that possesses potent antiemetic activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107007-99-8
Molecular Formula C18H25ClN4O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 348.87
Purity >99%
Molecular Weight (g/mol) 348.875
Chemical Name or Material Granisetron hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.